Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004119.2 | NP_004110.2 | 3, 4 | 460 | ||
NR_130706.1 | 3, 4 | 460 | ||||
XM_011535015.2 | XP_011533317.1 | 3, 4 | 459 | |||
XM_011535017.2 | XP_011533319.1 | 2, 3 | 283 | |||
XM_017020486.1 | XP_016875975.1 | 3, 4 | 455 | |||
XM_017020487.1 | XP_016875976.1 | 3, 4 | 441 | |||
XM_017020488.1 | XP_016875977.1 | 3, 4 | 432 | |||
XM_017020489.1 | XP_016875978.1 | 3, 4 | 451 | |||
GenBank mRNA | BC036028.1 | - | 3, 4 | 460 | ||
BC126350.1 | - | 3, 4 | 444 | 8992066 | ||
BC144039.1 | - | 3, 4 | 445 | 9052559 | ||
BC144040.1 | - | 3, 4 | 445 | 9052560 | ||
U02687.1 | - | 3, 4 | 435 | |||
Z26652.1 | - | 3, 4 | 378 |