Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccng1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012923.2 | NP_037055.1 | 2 | 326 | ||
GenBank mRNA | BC081852.1 | - | 2 | 297 | 7105829 | |
CB805791.1 | - | 2 | 347 | |||
FQ211577.1 | - | 2 | 378 | |||
FQ211975.1 | - | 2 | 378 | |||
FQ224502.1 | - | 2 | 287 | |||
X70871.1 | - | 2 | 296 |