Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012956.1 | NP_037088.1 | 2 | 224 | ||
GenBank mRNA | X15466.1 | - | 2 | 224 |