Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Icam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012967.1 | NP_037099.1 | 2 | 122 | ||
GenBank mRNA | BC081837.1 | - | 2 | 134 | 7122397 | |
D00913.1 | - | 2 | 122 | |||
FQ228241.1 | - | 2 | 142 |