Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MSX2 SIRNA Assays ›
Gene Symbol:
Msx2
Gene Name:
msh homeobox 2
Gene Aliases:
-
Chromosome Location:
Chr.17: 11683862 - 11689527 on Build Rnor_6.0
UniGene ID:
Rn.10414
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
48783
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Msx2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_012982.3
NP_037114.2
2
589
Back To Top
More Information
Panther Classification:
Molecular Function -
homeodomain transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
ossification
osteoblast differentiation
chondrocyte development
osteoblast development
outflow tract septum morphogenesis
outflow tract morphogenesis
epithelial to mesenchymal transition involved in endocardial cushion formation
endochondral bone growth
transcription from RNA polymerase II promoter
negative regulation of cell proliferation
anterior/posterior pattern specification
signal transduction involved in regulation of gene expression
embryonic limb morphogenesis
BMP signaling pathway
positive regulation of BMP signaling pathway
negative regulation of CREB transcription factor activity
embryonic forelimb morphogenesis
embryonic hindlimb morphogenesis
wound healing, spreading of epidermal cells
embryonic nail plate morphogenesis
wound healing
odontogenesis
embryonic digit morphogenesis
regulation of apoptotic process
positive regulation of apoptotic process
negative regulation of apoptotic process
negative regulation of fat cell differentiation
negative regulation of keratinocyte differentiation
positive regulation of osteoblast differentiation
negative regulation of transcription, DNA-templated
stem cell differentiation
cartilage development
positive regulation of catagen
bone trabecula formation
bone morphogenesis
frontal suture morphogenesis
branching involved in mammary gland duct morphogenesis
mammary gland epithelium development
BMP signaling pathway involved in heart development
enamel mineralization
cellular response to growth factor stimulus
cellular response to estradiol stimulus
activation of meiosis
negative regulation of transcription regulatory region DNA binding
positive regulation of mesenchymal cell apoptotic process
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, transcription factor binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
transcription cofactor activity
transcription factor binding
sequence-specific DNA binding
transcription regulatory region DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-gq622:80/100.66.78.111:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline