Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012983.2 | NP_037115.2 | 2 | 269 | ||
XM_008767949.1 | XP_008766171.1 | 2 | 2038 | |||
GenBank mRNA | BC087027.1 | - | 2 | 269 | 7106587 | |
X71997.1 | - | 2 | 319 |