Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkag1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013010.2 | NP_037142.1 | 3, 4 | 189 | ||
XM_006257331.2 | XP_006257393.1 | 3, 4 | 389 | |||
GenBank mRNA | BC085749.1 | - | 2, 3 | 175 | ||
BC097940.1 | - | 3, 4 | 189 | 7454478 | ||
U42413.1 | - | 2, 3 | 152 | |||
X95578.1 | - | 3, 4 | 209 |