Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St8sia3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013029.1 | NP_037161.1 | 1 | 165 | ||
GenBank mRNA | U55938.1 | - | 1 | 165 |