Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012926.1 | NP_037058.1 | 2 | 165 | ||
XM_006248365.2 | XP_006248427.1 | 2 | 1005 | |||
GenBank mRNA | AF010465.1 | - | 2 | 165 | ||
U05593.1 | - | 2 | 176 | |||
U10925.1 | - | 2 | 439 | |||
U88622.1 | - | 2 | 158 |