Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CK357656.1 | - | 2 | 242 | ||
FQ217150.1 | - | 2 | 272 | |||
FQ217655.1 | - | 2 | 274 | |||
FQ220032.1 | - | 531 | ||||
FQ220162.1 | - | 2 | 273 | |||
FQ221038.1 | - | 2 | 271 | |||
FQ221154.1 | - | 2 | 262 | |||
FQ221373.1 | - | 2 | 272 | |||
FQ222161.1 | - | 2 | 272 | |||
FQ222277.1 | - | 2 | 185 | |||
FQ222316.1 | - | 2 | 271 | |||
FQ222356.1 | - | 2 | 272 | |||
FQ222897.1 | - | 2 | 273 | |||
FQ223053.1 | - | 2 | 271 | |||
FQ224290.1 | - | 2 | 273 | |||
FQ224782.1 | - | 2 | 272 | |||
FQ228448.1 | - | 2 | 271 | |||
FQ228589.1 | - | 2 | 272 | |||
M55534.1 | - | 811 | ||||
S74229.1 | - | 2 | 235 | |||
S77138.1 | - | 2 | 249 | |||
S77142.1 | - | 2 | 234 | |||
X60351.1 | - | 2 | 241 |