Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgals4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012975.1 | NP_037107.1 | 3 | 239 | ||
XM_008759113.1 | XP_008757335.1 | 3 | 406 | |||
GenBank mRNA | M73553.1 | - | 3 | 239 |