Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012985.2 | NP_037117.1 | 4 | 372 | ||
GenBank mRNA | BC059148.1 | - | 4 | 235 | 6919202 | |
D86215.1 | - | 4 | 372 | |||
FQ212236.1 | - | 4 | 233 | |||
FQ228773.1 | - | 4 | 233 | |||
FQ229037.1 | - | 4 | 232 |