Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003742.2 | NP_003733.2 | 5 | 366 | ||
XM_006712817.3 | XP_006712880.1 | 5 | 478 | |||
XM_011512077.2 | XP_011510379.1 | 5 | 538 | |||
XM_011512078.2 | XP_011510380.1 | 5 | 538 | |||
XM_011512080.2 | XP_011510382.1 | 5 | 541 | |||
XM_017005165.1 | XP_016860654.1 | 5 | 540 | |||
GenBank mRNA | AF091582.1 | - | 5 | 366 | ||
AF136523.1 | - | 5 | 367 |