Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144070.1 | NP_001137542.1 | 5 | 660 | ||
NM_003786.3 | NP_003777.2 | 5 | 660 | |||
XM_005257763.3 | XP_005257820.1 | 5 | 674 | |||
XM_011525422.2 | XP_011523724.1 | 5 | 674 | |||
XM_011525423.1 | XP_011523725.1 | 5 | 674 | |||
XM_011525425.2 | XP_011523727.1 | 4 | 437 | |||
XM_017025265.1 | XP_016880754.1 | 1 | 473 | |||
XM_017025266.1 | XP_016880755.1 | 5 | 674 | |||
GenBank mRNA | AB010887.1 | - | 5 | 594 | ||
AB208954.1 | - | 5 | 600 | |||
AF009670.1 | - | 5 | 650 | |||
AF083552.1 | - | 5 | 597 | |||
AF085690.1 | - | 5 | 616 | |||
AF085691.1 | - | 5 | 616 | |||
AF085692.1 | - | 5 | 616 | |||
AF104943.1 | - | 5 | 584 | |||
BC046126.1 | - | 5 | 660 | |||
BC050370.1 | - | 5 | 585 | |||
BC058907.1 | - | 5 | 619 | |||
BC104952.1 | - | 5 | 612 | |||
BC137347.1 | - | 5 | 611 | 9021345 | ||
BC137348.1 | - | 5 | 611 | 9021346 | ||
Y17151.2 | - | 5 | 622 |