Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013028.1 | NP_037160.1 | 4, 5 | 428 | ||
GenBank mRNA | AJ002259.1 | - | 4, 5 | 428 |