Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRADD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320099.1 | NP_001307028.1 | 2 | 426 | ||
NM_001320100.1 | NP_001307029.1 | 2 | 216 | |||
NM_001320101.1 | NP_001307030.1 | 2 | 216 | |||
NM_003805.4 | NP_003796.1 | 2 | 216 | |||
NR_135147.1 | 2 | 216 | ||||
XM_017020142.1 | XP_016875631.1 | 2 | 245 | |||
XM_017020143.1 | XP_016875632.1 | 2 | 239 | |||
XM_017020144.1 | XP_016875633.1 | 2 | 239 | |||
XM_017020145.1 | XP_016875634.1 | 2 | 238 | |||
GenBank mRNA | AK294986.1 | - | 2 | 204 | ||
AK314861.1 | - | 2 | 196 | |||
BC017042.1 | - | 2 | 188 | 3849848 | ||
BC037905.1 | - | 2 | 430 | 5276510 | ||
BT009837.1 | - | 1 | 108 | |||
CR407643.1 | - | 1 | 108 | |||
HY113325.1 | - | 2 | 217 | |||
U79115.1 | - | 2 | 208 | |||
U84388.1 | - | 2 | 145 |