Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Id3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013058.2 | NP_037190.1 | 2 | 381 | ||
GenBank mRNA | AF000942.1 | - | 1 | 489 | ||
BC064658.1 | - | 2 | 381 | 5623701 | ||
D10864.1 | - | 2 | 358 | |||
FQ212997.1 | - | 2 | 403 | |||
FQ216267.1 | - | 2 | 403 | |||
FQ218468.1 | - | 2 | 401 | |||
FQ218958.1 | - | 2 | 394 | |||
FQ229332.1 | - | 2 | 403 |