Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013078.1 | NP_037210.1 | 2 | 256 | ||
GenBank mRNA | BC088158.1 | - | 2 | 222 | 7325875 | |
FQ211098.1 | - | 2 | 240 | |||
FQ219225.1 | - | 2 | 243 | |||
FQ219657.1 | - | 2 | 219 | |||
K00001.1 | - | 2 | 242 | |||
K03040.1 | - | 2 | 214 | |||
M11266.1 | - | 2 | 256 | |||
X01178.1 | - | 2 | 256 | |||
X01976.1 | - | 2 | 242 |