Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013091.1 | NP_037223.1 | 2 | 383 | ||
XM_006237334.2 | XP_006237396.1 | 2 | 456 | |||
GenBank mRNA | AF329976.1 | - | 2 | 382 | ||
AF329977.1 | - | 2 | 382 | |||
AF329978.1 | - | 2 | 382 | |||
AF329979.1 | - | 2 | 382 | |||
AF329980.1 | - | 2 | 382 | |||
AF329981.1 | - | 2 | 382 | |||
BC086413.1 | - | 2 | 403 | 7110094 | ||
M63122.1 | - | 2 | 383 |