Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igfbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013104.3 | NP_037236.1 | 2 | 417 | ||
GenBank mRNA | BC099742.1 | - | 2 | 429 | 7937967 | |
FQ220221.1 | - | 2 | 487 | |||
FQ220720.1 | - | 2 | 488 | |||
FQ220764.1 | - | 2 | 490 | |||
FQ230155.1 | - | 2 | 493 |