Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gckr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013120.2 | NP_037252.1 | 3, 4 | 458 | ||
GenBank mRNA | BC081843.1 | - | 3, 4 | 458 | 7096237 | |
KJ026952.1 | - | 3, 4 | 281 | |||
X68497.1 | - | 3, 4 | 302 |