Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKAR2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321982.1 | NP_001308911.1 | 1, 2 | 508 | ||
NM_001321983.1 | NP_001308912.1 | 1, 2 | 508 | |||
NM_001321989.1 | NP_001308918.1 | 1, 2 | 508 | |||
NM_004157.3 | NP_004148.1 | 1, 2 | 508 | |||
XM_005265315.3 | XP_005265372.1 | 1, 2 | 493 | |||
XM_011533942.2 | XP_011532244.1 | 1, 2 | 493 | |||
XM_011533943.1 | XP_011532245.1 | 1, 2 | 493 | |||
XM_017006815.1 | XP_016862304.1 | 1, 2 | 493 | |||
XM_017006816.1 | XP_016862305.1 | 1, 2 | 493 | |||
GenBank mRNA | AK293042.1 | - | 1, 2 | 462 | ||
BC002763.2 | - | 1, 2 | 486 | 3629579 | ||
BT007225.1 | - | 1, 2 | 254 | |||
CF994284.1 | - | 1, 2 | 522 | |||
DB040049.1 | - | 1, 2 | 490 | |||
KU178276.1 | - | 1, 2 | 254 | |||
KU178277.1 | - | 1, 2 | 254 | |||
X14968.1 | - | 1, 2 | 443 |