Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ywhah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013052.1 | NP_037184.1 | 2 | 365 | ||
GenBank mRNA | BC081825.1 | - | 2 | 390 | 7192904 | |
D17445.1 | - | 2 | 365 |