Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CREG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003851.2 | NP_003842.1 | 2, 3 | 546 | ||
GenBank mRNA | AF084523.1 | - | 2, 3 | 501 | ||
AK075375.1 | - | 2, 3 | 507 | |||
AK095456.1 | - | 3, 4 | 465 | |||
AK130006.1 | - | 3, 4 | 393 | |||
AK315497.1 | - | 2, 3 | 486 | |||
AY359071.1 | - | 2, 3 | 472 | |||
BC006786.1 | - | 2, 3 | 477 | 2900817 | ||
BC008628.1 | - | 2, 3 | 477 | 4339480 |