Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013091.1 | NP_037223.1 | 2, 3 | 438 | ||
GenBank mRNA | AF329976.1 | - | 2, 3 | 437 | ||
AF329977.1 | - | 2, 3 | 437 | |||
AF329978.1 | - | 2, 3 | 437 | |||
AF329979.1 | - | 2, 3 | 437 | |||
AF329980.1 | - | 2, 3 | 437 | |||
AF329981.1 | - | 2, 3 | 437 | |||
BC086413.1 | - | 2, 3 | 458 | 7110094 | ||
M63122.1 | - | 2, 3 | 438 |