Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cma1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013092.1 | NP_037224.1 | 4 | 393 | ||
GenBank mRNA | D38495.1 | - | 4 | 393 | ||
U67908.1 | - | 4 | 358 |