Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mc4r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013099.2 | NP_037231.1 | 1 | 563 | ||
GenBank mRNA | CB584477.1 | - | 1 | 563 | ||
U67863.1 | - | 1 | 302 |
Set Membership: |
G-protein coupled receptor |