Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013127.1 | NP_037259.1 | 6 | 729 | ||
GenBank mRNA | D29646.1 | - | 6 | 729 | ||
D30795.1 | - | 6 | 724 | |||
FQ212353.1 | - | 6 | 734 |