Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013066.1 | NP_037198.1 | 10, 11 | 5177 | ||
XM_008767203.1 | XP_008765425.1 | 10, 11 | 5357 | |||
XM_008767206.1 | XP_008765428.1 | 10, 11 | 5145 | |||
XM_008767208.1 | XP_008765430.1 | 8, 9 | 1163 | |||
GenBank mRNA | X17682.1 | - | 7, 8 | 1067 | ||
X51842.1 | - | 10, 11 | 5149 | |||
X54100.1 | - | 10, 11 | 5177 |