Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206897.1 | NP_001193826.1 | 5 | 832 | ||
NM_003888.3 | NP_003879.2 | 4 | 735 | |||
NM_170696.2 | NP_733797.1 | 4 | 735 | |||
NM_170697.2 | NP_733798.1 | 2 | 339 | |||
GenBank mRNA | AB015226.1 | - | 4 | 505 | ||
AB015227.1 | - | 2 | 268 | |||
AB015228.1 | - | 2 | 230 | |||
AK128709.1 | - | 4 | 1230 | |||
AK294981.1 | - | 4 | 435 | |||
AK303057.1 | - | 5 | 577 | |||
BC030589.2 | - | 4 | 493 | 4826743 | ||
BX443600.2 | - | 2 | 339 |