Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013147.1 | NP_037279.1 | 2 | 370 | ||
XM_008762873.1 | XP_008761095.1 | 2 | 1286 | |||
GenBank mRNA | AY112736.1 | - | 2 | 198 | ||
AY112737.1 | - | 2 | 198 | |||
AY112738.1 | - | 2 | 198 | |||
AY112739.1 | - | 2 | 198 | |||
AY112740.1 | - | 2 | 198 | |||
X62894.1 | - | 2 | 370 |