Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ucp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013167.2 | NP_037299.1 | 4 | 550 | ||
GenBank mRNA | AB006614.1 | - | 3 | 422 | ||
AF030163.1 | - | 3 | 437 | |||
AF035943.1 | - | 3 | 433 | |||
BC072546.1 | - | 4 | 550 | 7105743 | ||
U92069.1 | - | 4 | 557 |