Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkar1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013181.1 | NP_037313.1 | 2, 3 | 280 | ||
XM_008768311.1 | XP_008766533.1 | 2, 3 | 336 | |||
GenBank mRNA | M17086.1 | - | 2, 3 | 280 |