Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013226.2 | NP_037358.1 | 3 | 194 | ||
GenBank mRNA | BC061562.1 | - | 3 | 194 | 6921029 | |
BC082797.1 | - | 3 | 179 | 7301263 | ||
FQ209939.1 | - | 3 | 190 | |||
FQ210972.1 | - | 3 | 187 | |||
FQ211501.1 | - | 3 | 187 | |||
FQ211742.1 | - | 3 | 191 | |||
FQ212686.1 | - | 3 | 187 | |||
FQ216977.1 | - | 3 | 190 | |||
FQ216988.1 | - | 3 | 190 | |||
FQ217307.1 | - | 3 | 190 | |||
FQ217782.1 | - | 3 | 190 | |||
FQ217854.1 | - | 3 | 190 | |||
FQ219795.1 | - | 3 | 187 | |||
FQ220681.1 | - | 3 | 188 | |||
FQ221082.1 | - | 3 | 190 | |||
FQ221405.1 | - | 2 | 156 | |||
FQ221557.1 | - | 3 | 187 | |||
FQ221558.1 | - | 3 | 191 | |||
FQ221666.1 | - | 3 | 190 | |||
FQ221830.1 | - | 3 | 191 | |||
FQ221995.1 | - | 3 | 191 | |||
FQ222044.1 | - | 3 | 187 | |||
FQ222154.1 | - | 3 | 189 | |||
FQ222265.1 | - | 3 | 190 | |||
FQ222417.1 | - | 3 | 187 | |||
FQ222458.1 | - | 3 | 191 | |||
FQ222536.1 | - | 3 | 190 | |||
FQ222629.1 | - | 3 | 187 | |||
FQ222759.1 | - | 3 | 191 | |||
FQ222997.1 | - | 3 | 191 | |||
FQ223042.1 | - | 3 | 190 | |||
FQ223180.1 | - | 3 | 190 | |||
FQ223244.1 | - | 3 | 187 | |||
FQ224329.1 | - | 3 | 187 | |||
FQ224364.1 | - | 3 | 191 | |||
FQ224436.1 | - | 3 | 187 | |||
FQ228463.1 | - | 3 | 190 | |||
FQ228512.1 | - | 3 | 187 | |||
FQ228892.1 | - | 3 | 194 | |||
X06483.1 | - | 2 | 145 |