Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016993.1 | NP_058689.1 | 1 | 670 | ||
GenBank mRNA | AF149299.1 | - | 1 | 13 | ||
AF512835.1 | - | 1 | 436 | |||
L14680.1 | - | 1 | 670 | |||
U34964.1 | - | 1 | 436 |