Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF134 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003435.4 | NP_003426.3 | 3 | 1772 | ||
GenBank mRNA | AK291763.1 | - | 3 | 1741 | ||
BC011646.2 | - | 2 | 1631 | |||
BC021280.2 | - | 2 | 1616 | |||
BC042636.1 | - | 3 | 1731 | 4826076 | ||
BC112294.1 | - | 3 | 1532 | 8327762 | ||
BC113410.1 | - | 3 | 1529 | 8322462 | ||
U09412.1 | - | 3 | 1714 |