Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myh9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013194.1 | NP_037326.1 | 224 | |||
GenBank mRNA | U31463.1 | - | 224 |
Gene Symbol: LOC100911597 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750378.3 | XP_003750426.1 | 2 | 439 |