Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnai1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013145.1 | NP_037277.1 | 8 | 1168 | ||
GenBank mRNA | DQ120469.1 | - | 8 | 1042 | ||
DQ120470.1 | - | 8 | 1042 | |||
M17527.1 | - | 8 | 1168 |