Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc18a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013152.2 | NP_037284.2 | 5 | 832 | ||
XM_006252993.2 | XP_006253055.1 | 4 | 1032 | |||
XM_008771066.1 | XP_008769288.1 | 2 | 618 | |||
XM_008771067.1 | XP_008769289.1 | 4 | 1032 | |||
GenBank mRNA | AY168444.1 | - | 2 | 85 | ||
M97380.1 | - | 5 | 861 |