Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Maob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013198.1 | NP_037330.1 | 7, 8 | 839 | ||
GenBank mRNA | BC078886.1 | - | 7, 8 | 740 | ||
BC089814.1 | - | 7, 8 | 804 | 7323595 | ||
FQ210326.1 | - | 7, 8 | 900 | |||
M23601.1 | - | 7, 8 | 839 |