Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134709.1 | NP_001128181.1 | 10 | 1600 | ||
NM_003472.3 | NP_003463.1 | 11 | 1702 | |||
GenBank mRNA | AJ420539.1 | - | 3 | 540 | ||
BC035259.1 | - | 11 | 1674 | 5122743 | ||
BX641063.1 | - | 4 | 2805 | |||
X64229.1 | - | 11 | 1542 |