Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Add1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016990.2 | NP_058686.2 | 9 | 1194 | ||
XM_006251159.1 | XP_006251221.1 | 9 | 1308 | |||
XM_006251160.1 | XP_006251222.1 | 9 | 1308 | |||
XM_006251161.1 | XP_006251223.1 | 9 | 1308 | |||
XM_006251164.2 | XP_006251226.1 | 9 | 1308 | |||
XM_006251165.2 | XP_006251227.1 | 9 | 1308 | |||
XM_006251166.2 | XP_006251228.1 | 9 | 1308 | |||
XM_006251167.2 | XP_006251229.1 | 9 | 1308 | |||
XM_006251168.2 | XP_006251230.1 | 9 | 1308 | |||
XM_006251169.2 | XP_006251231.1 | 9 | 1308 | |||
XM_008770249.1 | XP_008768471.1 | 9 | 1308 | |||
XM_008770250.1 | XP_008768472.1 | 9 | 1308 | |||
GenBank mRNA | BC107657.1 | - | 9 | 1194 | 7935564 | |
Z49081.1 | - | 8 | 1133 | |||
Z49082.1 | - | 8 | 1133 |