Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017022.2 | NP_058718.2 | 10, 11 | 1447 | ||
XM_006255824.1 | XP_006255886.1 | 10, 11 | 1477 | |||
GenBank mRNA | BC131845.1 | - | 10, 11 | 1447 | 8366766 | |
U12309.1 | - | 10, 11 | 1321 |