Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113328.1 | NP_001106799.1 | 7 | 922 | ||
NM_001113329.1 | NP_001106800.1 | 8 | 837 | |||
NM_001113332.1 | NP_001106803.1 | 7 | 774 | |||
NM_001113334.1 | NP_001106805.1 | 7 | 611 | |||
NM_017032.1 | NP_058728.1 | 7 | 529 | |||
XM_008760721.1 | XP_008758943.1 | 7 | 1930 | |||
XM_008760722.1 | XP_008758944.1 | 2 | 7633 | |||
XM_008760723.1 | XP_008758945.1 | 3 | 273 | |||
GenBank mRNA | AF031373.1 | - | 7 | 922 | ||
AF536974.1 | - | 2 | 245 | |||
AF536979.1 | - | 8 | 837 | |||
AY388961.1 | - | 7 | 547 | |||
EF102484.1 | - | 7 | 774 | |||
EF121818.1 | - | 7 | 611 | |||
L27059.1 | - | 4 | 248 | |||
L27060.1 | - | 2 | 166 | |||
U09455.1 | - | 2 | 415 | |||
U09456.1 | - | 3 | 273 | |||
U09457.1 | - | 7 | 529 |