Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017034.1 | NP_058730.1 | 4 | 518 | ||
GenBank mRNA | X63675.1 | - | 4 | 518 |