Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppm1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017038.1 | NP_058734.1 | 2 | 186 | ||
XM_006240193.2 | XP_006240255.1 | 2 | 3489 | |||
XM_006240194.2 | XP_006240256.1 | 2 | 443 | |||
XM_006240195.2 | XP_006240257.1 | 2 | 207 | |||
GenBank mRNA | J04503.1 | - | 2 | 186 |