Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017059.2 | NP_058755.2 | 2, 3 | 181 | ||
GenBank mRNA | AF235993.1 | - | 1, 2 | 546 | ||
CB586216.1 | - | 2, 3 | 181 | |||
U49729.1 | - | 2, 3 | 92 |