Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017066.2 | NP_058762.1 | 2 | 168 | ||
XM_006236290.2 | XP_006236352.1 | 2 | 1552 | |||
XM_006236294.2 | XP_006236356.1 | 2 | 1148 | |||
GenBank mRNA | BC062013.1 | - | 2 | 168 | 5621277 | |
FQ213442.1 | - | 2 | 172 | |||
FQ223195.1 | - | 2 | 320 | |||
M55601.1 | - | 2 | 369 |