Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glul | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017073.3 | NP_058769.4 | 3 | 342 | ||
GenBank mRNA | BC072694.1 | - | 3 | 342 | 7103110 | |
FQ213256.1 | - | 3 | 353 | |||
FQ216093.1 | - | 3 | 347 | |||
FQ226911.1 | - | 3 | 365 | |||
FQ227474.1 | - | 3 | 354 | |||
FQ232853.1 | - | 3 | 403 | |||
FQ232963.1 | - | 3 | 354 | |||
FQ233770.1 | - | 3 | 354 | |||
FQ234289.1 | - | 3 | 355 | |||
M29579.1 | - | 3 | 353 | |||
M91652.1 | - | 3 | 351 | |||
X07921.1 | - | 3 | 353 |