Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Umod | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017082.1 | NP_058778.1 | 2 | 130 | ||
XM_006230107.2 | XP_006230169.1 | 3 | 252 | |||
GenBank mRNA | BC081814.1 | - | 2 | 136 | 7109450 | |
M63510.1 | - | 2 | 130 | |||
S75960.1 | - | 2 | 121 |